Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPRC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584352
Description
Immunogen sequence: LKPEGVTEAK HPAAVRLQEG VHGPSRVHVG SGDHDYCVRS RTPPKKMPAL VIPEVGSRWN VKRHQDITIK PVLSLGPAAP PPPCIAASRE PLDHRTSSEQ ADPSAP.
The protein encoded by this gene is similar to PPAR-gamma coactivator 1 (PPARGC1/PGC-1), a protein that can activate mitochondrial biogenesis in part through a direct interaction with nuclear respiratory factor 1 (NRF1). This protein has been shown to interact with NRF1. It is thought to be a functional relative of PPARGC1 that activates mitochondrial biogenesis through NRF1 in response to proliferative signals.
Specifications
PPRC1 | |
Polyclonal | |
Unconjugated | |
PPRC1 | |
A930018M19; BC013720; KIAA0595; mKIAA0595; peroxisome proliferative activated receptor, gamma, coactivator-related 1; peroxisome proliferator-activated receptor gamma coactivator-related protein 1; peroxisome proliferator-activated receptor gamma, coactivator-related 1; PGC-1-related coactivator; PPRC1; PRC; RP11-302K17.6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23082 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VV67 | |
PPRC1 | |
Recombinant protein corresponding to Human PPRC1. Recombinant protein control fragment (Product #RP-106788). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction