Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PPT
GSA_VAGREENER_CHOICE

Catalog No. 142610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-261-0 10 mg
14-265-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-261-0 Supplier Tocris Bioscience™ Supplier No. 1426/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Subtype-selective ERα agonist

Potent, subtype-selective estrogen receptor agonist (EC50 ∽ 200 pM); displays 410-fold selectivity for ERα over ERβ. Prevents ovariectomy-induced weight gain and loss of bone mineral density, and induces gene expression in the hypothalamus following systemic administration in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 263717-53-9
Molecular Formula C24H22N2O3

Specifications

Quantity 10 mg
Formula Weight 386.45
Purity >99%
Molecular Weight (g/mol) 386.45
Chemical Name or Material PPT
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.