Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPT1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300363MFV610
Description
PPT1 Polyclonal antibody specifically detects PPT1 in Human samples. It is validated for ELISASpecifications
PPT1 | |
Polyclonal | |
ELISA | |
ceroid-palmitoyl-palmitoyl-protein thioesterase 1, CLN1EC 3.1.2.22, INCL, Palmitoyl-protein hydrolase 1, palmitoyl-protein thioesterase 1, PPT, PPT-1 | |
Produced in rabbits immunized with purified, recombinant Human PPT1 (Accession#: P50897-1; Met1-Gly306) | |
0.1 mL | |
Neuroscience | |
5538 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction