Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110874
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110874 Supplier Invitrogen™ Supplier No. PA5110874
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KFVMVKFLND SIVDPVDSEW FGFYRSGQAK ETIPLQETSL YTQDRLGLKE MDNAGQLVFL ATEGDHLQLS EEWFY Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 87%.

The protein encoded by this gene is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. The encoded enzyme removes thioester-linked fatty acyl groups such as palmitate from cysteine residues. Defects in this gene are a cause of infantile neuronal ceroid lipofuscinosis 1 (CLN1, or INCL) and neuronal ceroid lipofuscinosis 4 (CLN4). Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ppt1
Gene Accession No. P50897
Gene Alias 3.1.2.22; 9530043G02Rik; AA960502; C77813; ceroid-palmitoyl-palmitoyl-protein thioesterase 1; CLN1; D4Ertd184e; INCL; Palmitoyl-protein hydrolase 1; palmitoyl-protein thioesterase; palmitoyl-protein thioesterase 1; palmitoyl-protein thioesterase 1 (ceroid-lipofuscinosis, neuronal 1, infantile); PPT; Ppt1; PPT-1; QnpA-18851; unnamed protein product
Gene Symbols Ppt1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human PPT1. Recombinant protein control fragment (Product #RP-92871).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5538
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.