Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586510
Description
Purity is > 95% by SDS-PAGE.
The protein encoded by this gene is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. The encoded enzyme removes thioester-linked fatty acyl groups such as palmitate from cysteine residues. Defects in this gene are a cause of infantile neuronal ceroid lipofuscinosis 1 (CLN1, or INCL) and neuronal ceroid lipofuscinosis 4 (CLN4). Two transcript variants encoding different isoforms have been found for this gene.
Specifications
PPT1 | |
Polyclonal | |
Unconjugated | |
Ppt1 | |
3.1.2.22; 9530043G02Rik; AA960502; C77813; ceroid-palmitoyl-palmitoyl-protein thioesterase 1; CLN1; D4Ertd184e; INCL; Palmitoyl-protein hydrolase 1; palmitoyl-protein thioesterase; palmitoyl-protein thioesterase 1; palmitoyl-protein thioesterase 1 (ceroid-lipofuscinosis, neuronal 1, infantile); PPT; Ppt1; PPT-1; QnpA-18851; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19063, 29411, 5538 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O88531, P45479, P50897 | |
Ppt1 | |
Synthetic peptide, corresponding to amino acids 50-100 of Human CLN1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction