Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548700
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548700 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548700 Supplier Invitrogen™ Supplier No. PA548700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPT2 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the palmitoyl-protein thioesterase family. The encoded glycosylated lysosomal protein has palmitoyl-CoA hydrolase activity in vitro, but does not hydrolyze palmitate from cysteine residues in proteins. Three transcript variants encoding different isoforms have been described for this gene, with one of the isoforms being inactive.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PPT2
Gene Accession No. Q9UMR5
Gene Alias 0610007M19Rik; AA672937; C6orf8; G14; Lysosomal thioesterase PPT2; palmitoyl-protein hydrolase 2; palmitoyl-protein thioesterase 2; PPT2; PPT-2; S-thioesterase G14; VE-statin2
Gene Symbols PPT2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 183-212 amino acids from the Central region of human PPT2.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9374
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.