Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588446 Supplier Invitrogen™ Supplier No. PA588446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: APAPHRASYK PVIVVHGLFD SSYSFRHLLE YINETHPGTV VTVLDLFDGR ESLRPLWEQV QGFREAVVPI MAKAPQGVHL ICYSQGGLVC RALLSVMDDH NVDSFISLSS PQMGQYGDTD YLKWLFPTSM RSNLYRICYS PWGQEFSICN YWHDPHHDDL YLNASSFLAL INGERDHPNA TVWRKNFLRV GHLVLIGGPD DGVITPWQSS FFGFYDANET VLEMEEQLVY LRDSFGLKTL LARGAIVRCP MAGISHTAWH SNRTLYETCI EPWLS; Positive Samples: Mouse liver; Cellular Location: Lysosome.

This gene encodes a member of the palmitoyl-protein thioesterase family. The encoded glycosylated lysosomal protein has palmitoyl-CoA hydrolase activity in vitro, but does not hydrolyze palmitate from cysteine residues in proteins. Three transcript variants encoding different isoforms have been described for this gene, with one of the isoforms being inactive.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PPT2
Gene Accession No. O35448
Gene Alias 0610007M19Rik; AA672937; C6orf8; G14; Lysosomal thioesterase PPT2; palmitoyl-protein hydrolase 2; palmitoyl-protein thioesterase 2; PPT2; PPT-2; S-thioesterase G14; VE-statin2
Gene Symbols PPT2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 28-302 of human PPT2 (NP_0051464).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54397
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.