Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPTC7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559223
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559223 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559223 Supplier Invitrogen™ Supplier No. PA559223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGDIILTATD GLFDNMPDYM ILQELKKLKN SNYESIQQTA RSIAEQAHEL AYDPNYMSPF AQFACDNGLN VRGGKPDDIT VLLSIVAEY Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PPTC7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPTC7
Gene Accession No. Q8NI37
Gene Alias 9130017A15Rik; AA672638; AI848390; PPTC7; Protein phosphatase PTC7 homolog; PTC7 protein phosphatase homolog; PTC7 protein phosphatase homolog (S. cerevisiae); RGD1310383; TAPP2C; TA-PP2C; T-cell activation protein phosphatase 2C; T-cell activation protein phosphatase 2C-like
Gene Symbols PPTC7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPTC7. Recombinant protein control fragment (Product #RP-97886).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160760
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.