Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPTC7 Polyclonal Antibody

Supplier: Thermo Scientific PA5114290
Description
Specifications
| PPTC7 | |
| Polyclonal | |
| Unconjugated | |
| PPTC7 | |
| 9130017A15Rik; AA672638; AI848390; PPTC7; Protein phosphatase PTC7 homolog; PTC7 protein phosphatase homolog; PTC7 protein phosphatase homolog (S. cerevisiae); RGD1310383; TAPP2C; TA-PP2C; T-cell activation protein phosphatase 2C; T-cell activation protein phosphatase 2C-like | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 160760 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NI37 | |
| PPTC7 | |
| Synthetic peptide directed towards the C-terminal region of Human PPTC7 (aa 230-279). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction