Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPWD1 Antibody (OTI1B8), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273590FR
Description
PPWD1 Monoclonal specifically detects PPWD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PPWD1 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
PPWD1 | |
Full length recombinant protein of human PPWD1 (NP_056157) produced in HEK293T cells. | |
0.1 mL | |
23398.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1B8 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 5.2.1.8, KIAA0073Spliceosome-associated cyclophilin, peptidylprolyl isomerase domain and WD repeat containing 1, peptidylprolyl isomerase domain and WD repeat-containing protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction