Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPWD1 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502076S


Description
PPWD1 Monoclonal antibody specifically detects PPWD1 in Canine, Human, Rat samples. It is validated for Western BlotSpecifications
PPWD1 | |
Monoclonal | |
0.52 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q16740 | |
Mouse | |
Affinity chromatography | |
RUO | |
301117, 611738, 8192 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI2B1 | |
Unconjugated | |
CLPP | |
4632422M10Rik; A330090G21Rik; KIAA0073; peptidylprolyl isomerase domain and WD repeat containing 1; peptidylprolyl isomerase domain and WD repeat-containing protein 1; PPWD1; RGD1310204; spliceosome-associated cyclophilin | |
Full length human recombinant protein of human CLPP produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction