Learn More
PPWD1 Mouse anti-Human, Clone: OTI2B1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502076

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Putative peptidylprolyl isomerase (PPIase). PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides. May be involved in pre-mRNA splicing.Specifications
PPWD1 | |
Monoclonal | |
Unconjugated | |
PPWD1 | |
peptidylprolyl isomerase domain and WD repeat containing 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23398 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2B1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96BP3 | |
PPWD1 | |
Full length human recombit protein of human PPWD1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.