Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPWD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553982 Supplier Invitrogen™ Supplier No. PA553982
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GALFCSVGDD KAMKVFDVVN FDMINMLKLG YFPGQCEWIY CPGDAISSVA ASEKSTGKIF IYDGRGDNQP LHIFDKLHTS PLTQIRLNPV YKAVVS Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

PPIase that catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and may therefore assist protein folding (PubMed:20676357). May be involved in pre-mRNA splicing (PubMed:11991638). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPWD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPWD1
Gene Accession No. Q96BP3
Gene Alias 4632422M10Rik; A330090G21Rik; KIAA0073; peptidylprolyl isomerase domain and WD repeat containing 1; peptidylprolyl isomerase domain and WD repeat-containing protein 1; PPWD1; RGD1310204; spliceosome-associated cyclophilin
Gene Symbols PPWD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPWD1. Recombinant protein control fragment (Product #RP-91728).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23398
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.