Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPY Monoclonal Antibody (2G5D12)
GREENER_CHOICE

Catalog No. PIMA517157
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517157 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517157 Supplier Invitrogen™ Supplier No. MA517157
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17157 targets PPY in FACS, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17157 immunogen is purified recombinant fragment of human PPY (amino acids: 1-95) expressed in E. Coli. MA5-17157 detects PPY which has a predicted molecular weight of approximately 10.4kDa.

The PPY gene encodes pancreatic polypeptide, a peptide hormone involved in the regulation of appetite and energy balance. The origin of the PPY gene is linked to a tandem duplication of the PYY gene, occurring concomitantly with the emergence of tetrapods, leading to the distinct PPY function within the neuropeptide Y family. In primates and ungulates, a gene duplication event resulted in the formation of a PYY2-PPY2 gene cluster, highlighting the evolutionary complexity of these genes. The PPY gene product is secreted by gamma-cells in the pancreas, localized primarily in the islet periphery, and plays a crucial role in metabolic homeostasis by interacting with receptors in the central nervous system to modulate satiety signals postprandially. PPY's function is linked to its inhibition of AGRP/NPY neurons which decreases their suppressive effect on POMC neurons, enhancing POMC activity and contributing to appetite suppression. This intricate regulation underscores the importance of PPY in dietary intake and metabolic disorders, with ongoing research exploring its potential therapeutic applications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPY
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2G5D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PPY
Gene Accession No. P01298
Gene Alias AV061413; C-terminal peptide; HGNC:9327; IGSPP; IOPPP; Obinepitide; panceatic polypeptide; Pancreatic hormone; Pancreatic icosapeptide; Pancreatic polypeptide; pancreatic polypeptide Y; pancreatic prohormone; Pancreatic prohormone percursor; pancreatic prohormone precursor; PH; PI; PNP; PP; PP1; PPA1; PPase; Ppy; prepro-PP (prepropancreatic polypeptide); SID6-8061
Gene Symbols PPY
Host Species Mouse
Immunogen Purified recombinant fragment of human PPY (amino acids: 1-95) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5539
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.