Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PQ 401
GREENER_CHOICE

Catalog No. 276810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-681-0 10 mg
27-685-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-681-0 Supplier Tocris Bioscience™ Supplier No. 2768/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

IGF1R inhibitor

Insulin-like growth factor receptor (IGF1R) inhibitor. Suppresses IGF-stimulated IGF-IR autophosphorylation with an IC50 value of 12 μM. Inhibits growth of MCF-7 breast cancer cells in vitro and in vivo.

Chemical Identifiers

CAS 196868-63-0
Molecular Formula C18H16ClN3O2

Specifications

Quantity 10 mg
Formula Weight 341.79
Purity >99%
Molecular Weight (g/mol) 341.80
Chemical Name or Material PQ 401
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.