Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PQBP1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP147270AF488
Description
PQBP1 Polyclonal antibody specifically detects PQBP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
PQBP1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
38 kDa nuclear protein containing a WW domain, MRX55, MRXS3, MRXS8, Npw38, NPW38mental retardation, X-linked 55, nuclear protein containing WW domain 38 kD, polyglutamine binding protein 1, Polyglutamine tract-binding protein 1, polyglutamine-binding protein 1, PQBP-1, RENS1, SHS, Sutherland-Haan X-linked mental retardation syndrome | |
The immunogen for this product maps to a region between residue 150 and 200 of human Polyglutamine Binding Protein 1 using the numbering given in entry NP_005701.1 (GeneID 10084). | |
0.1 mL | |
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
10084 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction