Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PQBP1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB261854 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB261854 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB261854 Supplier Novus Biologicals Supplier No. NBP147270H
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PQBP1 Polyclonal antibody specifically detects PQBP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen PQBP1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 38 kDa nuclear protein containing a WW domain, MRX55, MRXS3, MRXS8, Npw38, NPW38mental retardation, X-linked 55, nuclear protein containing WW domain 38 kD, polyglutamine binding protein 1, Polyglutamine tract-binding protein 1, polyglutamine-binding protein 1, PQBP-1, RENS1, SHS, Sutherland-Haan X-linked mental retardation syndrome
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 150 and 200 of human Polyglutamine Binding Protein 1 using the numbering given in entry NP_005701.1 (GeneID 10084).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10084
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.