Learn More
Invitrogen™ PQBP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551651
Description
Immunogen sequence: PVALQTRLAK RGILKHLEPE PEEEIIAEDY DDDPVDYEAT RLEGLPPSWY KVFDPSCGLP YYWNADTDLV SWLSPHDPNS VVTKSAKKLR SSNADAEEKL DRSHDKSDRG HDKSDRSHEK LDRGHDKSDR GHDKSDRDRE RGYDKVD Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
PQBP1 is a nuclear polyglutamine-binding protein that contains a WW domain. Mutations in this gene are associated with X-linked mental retardation.
Specifications
PQBP1 | |
Polyclonal | |
Unconjugated | |
Pqbp1 | |
38 kDa nuclear protein containing a WW domain; JM26; MGC129091 protein; MRX2; MRX55; MRXS3; MRXS8; NPW38; polyglutamine binding protein 1; polyglutamine tract-binding protein 1; Polyglutamine-binding protein 1; PQBP1; PQBP-1; RENS1; scurfy candidate 2; Sfc2; SHS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10084 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60828 | |
Pqbp1 | |
Recombinant protein corresponding to Human PQBP1. Recombinant protein control fragment (Product #RP-92128). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.