Learn More
PQBP1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302802A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Gorilla
PQBP1 is a nuclear polyglutamine-binding protein that contains a WW domain. Mutations in this gene are associated with X-linked mental retardation.Specifications
PQBP1 | |
Polyclonal | |
Unconjugated | |
PQBP1 | |
38 kDa nuclear protein containing a WW domain, mental retardation, X-linked 2 (non-dysmorphic), mental retardation, X-linked 55, MRX2, MRX55, MRXS3, MRXS8, NPW38, polyglutamine tract-binding protein 1, PQBP-1, RENS1, SHS, Sutherland-Haan X-linked mental retardation syndrome | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10084, 54633 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O60828, Q91VJ5 | |
PQBP1 | |
Between 215 and 265 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.