Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PQLC2L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570989
Description
This target displays homology in the following species: Human: 100%.
PQLC2L is a protein coding gene.
Specifications
PQLC2L | |
Polyclonal | |
Unconjugated | |
PQLC2L | |
C3orf55; PQ loop repeat containing 2 like; PQ loop repeat containing 2-like; PQLC2L; PQ-loop repeat-containing protein 2-like; putative uncharacterized protein PQLC2L; Putative uncharacterized protein SLC66A1L; SLC66A1L; Solute carrier family 66 member 1-like | |
Rabbit | |
Affinity Chromatography | |
RUO | |
152078 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A1A4F0 | |
PQLC2L | |
synthetic peptide directed towards the N-terminal region of human PQLC2L. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction