Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoefer™ PR 648 Slot Blot Blotting Manifold

Catalog No. 45002241 Shop All Cytiva Products
Change view
Click to view available options
Type:
Slot Blot Manifold
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
45-002-241 Slot Blot Manifold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-241 Supplier Hoefer™ Supplier No. 80609558

missing translation for 'validMainframeMsg'

Includes

Top block, Membrane support block with O-ring, Bottom block with connector, QuickFit connector with O-ring, Six clamping screws, Membrane cutting template

Provides a simple, convenient method for screening recombinant clones, DNA, RNA and proteins.

Forty-eight slots in a 4 × 12 grid empty into a single lower chamber which has a fitting for attaching a vacuum pump or aspirator to draw samples through the slot. Each slot is 6 mm long, 0.80 mm wide, and exposes 4.8 mm2 of membrane.

  • Raised lip around each slot creates an effective seal and prevents cross-contamination between wells
  • Alphanumeric grid facilitates accurate sample application
  • Alignment pins ensure quick and easy assembly
  • Membrane blotted in the Slot Blot, or autoradiograms made from the membranes, are easily quantified by densitometry
  • Sturdy acrylic construction

TRUSTED_SUSTAINABILITY

Specifications

Type Slot Blot Manifold
Includes Top block, membrane support block with O-ring, bottom block with connector, QuickFit™ connector with O-ring, 6 clamping screws, membrane cutting template
Material Acrylic
No. of Slots 48
Temperature (English) Maximum 113°F
Temperature (Metric) Maximum 45°C
Volume (Metric) Sample 1 mL
Length (English) 3.14 in.
Length (Metric) 8 cm
Width (English) 6.1 in.
Width (Metric) 15.5 cm
Height (Metric) 12 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.