Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735109 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5109 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5109 Supplier Proteintech Group Inc Supplier No. 258711AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The progesterone receptor is a member of the Steroid Hormone Receptor Superfamily. It exists in two distinct isoforms in human cells. PR-B is the transcriptionally active form and is responsible for activating genes for the maintainence of the endometrium, maintece of pregcy, and inhibition of ovulation. PR-A is identical to PR-B except for a 165 amino acid deletion at the N-terminus. This deletion exposes a 140 amino acid inhibitory domain (ID) that acts as a repressor of steroid hormone transcriptional activity. Phospholabeling studies have shown that PR is phosphorylated in at least 9 different locations in response to progestins, which suggests that PR affects many different cellular pathways.

Specifications

Antigen PR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PGR
Gene Accession No. P06401, Q00175
Gene Alias NR3C3, PGR, PR, progesterone receptor
Gene Symbols Pgr
Host Species Rabbit
Immunogen PR Fusion Protein Ag22986
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18667, 5241
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.