Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PR3 Monoclonal Antibody (3B4)
GREENER_CHOICE

Catalog No. PIMA515540
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515540 Supplier Invitrogen™ Supplier No. MA515540
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15540 targets Proteinase 3 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15540 immunogen is purified recombinant fragment of human Proteinase 3 expressed in E. Coli. MA5-15540 detects Proteinase 3 which has a predicted molecular weight of approximately 28kDa.

Serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) (PubMed:3198760, PubMed:2033050, PubMed:28240246). By cleaving and activating receptor F2RL1/PAR-2, enhances endothelial cell barrier function and thus vascular integrity during neutrophil transendothelial migration (PubMed:23202369). May play a role in neutrophil transendothelial migration, probably when associated with CD177 (PubMed:22266279). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PR3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3B4
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene PRTN3
Gene Accession No. P24158
Gene Alias ACPA; AGP7; azurophil granule protein 7; CANCA; C-ANCA; C-ANCA antigen; leukocyte proteinase 3; MBN; MBT; mPR3; Myeloblastin; neutrophil proteinase 4; NP4; NP-4; P29; PR3; PR-3; Proteinase 3; PRTN3; serine proteinase, neutrophil; wegener autoantigen; Wegener granulomatosis autoantigen
Gene Symbols PRTN3
Host Species Mouse
Immunogen Purified recombinant fragment of human Proteinase 3 expressed in E. Coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5657
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.