Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PR3 Monoclonal Antibody (MCPR3-2), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA511945
Description
MA5-11945 targets Proteinase 3 in IF, FACS and WB applications and shows reactivity with mouse and human samples. The MA5-11945 immunogen is hMC-1/PR3 cell granule extract.
Serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) (PubMed:3198760, PubMed:2033050, PubMed:28240246). By cleaving and activating receptor F2RL1/PAR-2, enhances endothelial cell barrier function and thus vascular integrity during neutrophil transendothelial migration (PubMed:23202369). May play a role in neutrophil transendothelial migration, probably when associated with CD177 (PubMed:22266279). [UniProt]
Specifications
PR3 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P24158, Q61096 | |
PRTN3 | |
HMC-1/PR3 cell granule extract | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry, Western Blot | |
MCPR3-2 | |
Unconjugated | |
PRTN3 | |
ACPA; AGP7; azurophil granule protein 7; CANCA; C-ANCA; C-ANCA antigen; leukocyte proteinase 3; MBN; MBT; mPR3; Myeloblastin; neutrophil proteinase 4; NP4; NP-4; P29; PR3; PR-3; Proteinase 3; PRTN3; serine proteinase, neutrophil; wegener autoantigen; Wegener granulomatosis autoantigen | |
Mouse | |
Protein G | |
RUO | |
19152, 5657 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction