Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PR3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110224
Description
Immunogen sequence: IVGGHEAQPH SRPYMASLQM RGNPGSHFCG GTLIHPSFVL TAAHCLRDIP QRLVNVVLGA HNVRTQEPTQ QHFSVAQVFL NNYDAENKLN DVLLIQLSSP ANLSASVATV QLPQQDQPVP HGTQCLAMGW GRVGAHDPPA QVL.
Serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) (PubMed:3198760, PubMed:2033050, PubMed:28240246). By cleaving and activating receptor F2RL1/PAR-2, enhances endothelial cell barrier function and thus vascular integrity during neutrophil transendothelial migration (PubMed:23202369). May play a role in neutrophil transendothelial migration, probably when associated with CD177 (PubMed:22266279). [UniProt].
Specifications
PR3 | |
Polyclonal | |
Unconjugated | |
PRTN3 | |
ACPA; AGP7; azurophil granule protein 7; CANCA; C-ANCA; C-ANCA antigen; leukocyte proteinase 3; MBN; MBT; mPR3; Myeloblastin; neutrophil proteinase 4; NP4; NP-4; P29; PR3; PR-3; Proteinase 3; PRTN3; serine proteinase, neutrophil; wegener autoantigen; Wegener granulomatosis autoantigen | |
Rabbit | |
Affinity Chromatography | |
RUO | |
314615 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.75 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
0 | |
PRTN3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 28-170 of human Proteinase 3/PR3 (NP_0027683). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction