Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA582223
Description
Immunogen sequence: PTQQHFSVAQ VFLNNYDAEN KLNDILLIQL SSPANLSASV ATVQLPQQDQ PVPHGTQCLA MGWGRVGAHD PPAQVLQELN VTVVTFFCRP HNICTFVPRR KAGICFGDSG GPLICDGI
Proteinase 3 plays a role in neutrophil response to inflammation.
Specifications
PR3 | |
Polyclonal | |
Unconjugated | |
PRTN3 | |
ACPA, AGP7, C-ANCA, MBT, P29, PR-3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5657 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P24158 | |
PRTN3 | |
Recombit protein corresponding to Human PRTN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction