Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PR3 Recombinant Rabbit Monoclonal Antibody (3I3M1)
GREENER_CHOICE

Catalog No. PIMA542734
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542734 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542734 Supplier Invitrogen™ Supplier No. MA542734
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: Mouse liver. Immunogen sequence: MAHRPPSPAL ASVLLALLLS GAARAAEIVG GHEAQPHSRP YMASLQMRGN PGSHFCGGTL IHPSFVLTAA HCLRDIPQRL VNVVLGAHNV RTQEPTQQHF.

Serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) (PubMed:3198760, PubMed:2033050, PubMed:28240246). By cleaving and activating receptor F2RL1/PAR-2, enhances endothelial cell barrier function and thus vascular integrity during neutrophil transendothelial migration (PubMed:23202369). May play a role in neutrophil transendothelial migration, probably when associated with CD177 (PubMed:22266279). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PR3
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Recombinant Monoclonal
Clone 3I3M1
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRTN3
Gene Accession No. P24158
Gene Alias ACPA; AGP7; azurophil granule protein 7; CANCA; C-ANCA; C-ANCA antigen; leukocyte proteinase 3; MBN; MBT; mPR3; Myeloblastin; neutrophil proteinase 4; NP4; NP-4; P29; PR3; PR-3; Proteinase 3; PRTN3; serine proteinase, neutrophil; wegener autoantigen; Wegener granulomatosis autoantigen
Gene Symbols PRTN3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Proteinase 3/PR3 (P24158).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5657
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.