Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PR38A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562730 Supplier Invitrogen™ Supplier No. PA562730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEDFKYVRML GALYMRLTGT AIDCYKYLEP LYNDYRKIKS QNRNGEFELM HVDEFIDELL HSERVCDIIL PRLQKRYVL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 100%.

PRPF38A is a protein coding gene involved in pre-mRNA splicing as a component of the spliceosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PR38A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRPF38A
Gene Accession No. Q8NAV1
Gene Alias 2410002M20Rik; pre-mRNA processing factor 38A; pre-mRNA-splicing factor 38A; Prp38; PRP38 pre-mRNA processing factor 38 (yeast) domain containing A; PRP38 pre-mRNA processing factor 38 domain containing A; Prpf38a; RP5-965L7.1
Gene Symbols PRPF38A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PR38A. Recombinant protein control fragment (Product #RP-94221).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84950
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.