Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAC Antibody, HRP , Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297053H
Description
PRAC Polyclonal antibody specifically detects PRAC in Human samples. It is validated for ELISASpecifications
PRAC | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
HRP | |
C17orf92, chromosome 17 open reading frame 92, MGC32520, PRAC1, prostate cancer susceptibility candidate, Prostate, rectum and colon expressed gene protein, small nuclear protein PRAC | |
Produced in rabbits immunized with purified, recombinant Human PRAC (NP_115767.1; Met1-Phe57). | |
0.1 mL | |
Cancer, Cell Biology, Tumor Suppressors | |
84366 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction