Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRAC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA146237
Description
PRAC Polyclonal Antibody for Western Blot, IHC (F), IHC (P), ELISA
PRAC is a novel gene that is expressed in human prostate, prostate cancer, rectum and distal colon. The nuclear localization, identification of potential phosphorylation sites, and possible cotranscription with the Hoxb-13 gene suggest that PRAC may have a regulatory role in the nucleus.
Specifications
PRAC | |
Polyclonal | |
Unconjugated | |
PRAC1 | |
C17orf92; MGC32520; PRAC; PRAC1; prostate cancer susceptibility candidate 1; prostate cancer susceptibility candidate protein 1; Prostate, rectum and colon expressed gene protein; small nuclear protein PRAC; small nuclear protein PRAC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84366 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q96KF2 | |
PRAC1 | |
Full length protein (human). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction