Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561612
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561612 Supplier Invitrogen™ Supplier No. PA561612
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLCAHFSDQG PAHLTTSKSA FLSNKKTSTL KHLLGETRSD GSACNSGISG GRGRKIP Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 32%.

PRAC is a novel gene that is expressed in human prostate, prostate cancer, rectum and distal colon. The nuclear localization, identification of potential phosphorylation sites, and possible cotranscription with the Hoxb-13 gene suggest that PRAC may have a regulatory role in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRAC1
Gene Accession No. Q96KF2
Gene Alias C17orf92; MGC32520; PRAC; PRAC1; prostate cancer susceptibility candidate 1; prostate cancer susceptibility candidate protein 1; Prostate, rectum and colon expressed gene protein; small nuclear protein PRAC; small nuclear protein PRAC1
Gene Symbols PRAC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRAC. Recombinant protein control fragment (Product #RP-93342).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84366
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.