Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14260
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14260 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14260 Supplier Invitrogen™ Supplier No. PA14260
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4260 detects the PRAC protein in human samples. PA1-4260 has been successfully used in Western blot procedures. PA1-4260 immunizing peptide corresponds to amino acid residues 4-19 of the human PRAC protein.

PRAC is a novel gene that is expressed in human prostate, prostate cancer, rectum and distal colon. The nuclear localization, identification of potential phosphorylation sites, and possible cotranscription with the Hoxb-13 gene suggest that PRAC may have a regulatory role in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAC
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PRAC1
Gene Accession No. Q96KF2
Gene Alias C17orf92; MGC32520; PRAC; PRAC1; prostate cancer susceptibility candidate 1; prostate cancer susceptibility candidate protein 1; Prostate, rectum and colon expressed gene protein; small nuclear protein PRAC; small nuclear protein PRAC1
Gene Symbols PRAC1
Host Species Rabbit
Immunogen Synthetic peptide: A(4) H F S D Q G P A H L T T S K S(19).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84366
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.