Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAK Antibody (OTI5E1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273594H
Description
PRAK Monoclonal specifically detects PRAK in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PRAK | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
MAPKAPK5 | |
Recombinant protein fragment corresponding to amino acids 197-471 of human PRAK (NP_003659) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot | |
OTI5E1 | |
Western Blot | |
EC 2.7.11, MAP kinase-activated protein kinase 5, MAPK-activated protein kinase 5, MAPKAP kinase 5, MAPKAPK-5, mitogen-activated protein kinase-activated protein kinase 5, p38-regulated/activated protein kinase, PRAKEC 2.7.11.1 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
8550.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction