Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528616 Supplier Invitrogen™ Supplier No. PA528616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Mouse (98%), Rat (96%), Zebrafish (92%), Pig (98%), Chicken (97%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

MAPKAPK5 (PRAK) is activated in response to cellular stress and pro-inflammatory cytokines. Activity of this kinase is regulated by p38 alpha and p38 beta. The protein encoded by PRAK is a member of the serine/threonine kinase family. In response to cellular stress and proinflammatory cytokines, this kinase is activated through its phosphorylation by MAP kinases including MAPK1/ERK, MAPK14/p38-alpha, and MAPK11/p38-beta. In vitro, this kinase phosphorylates heat shock protein HSP27 at its physiologically relevant sites. Two alternately spliced transcript variants of PRAK encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAK
Applications Western Blot
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene MAPKAPK5
Gene Accession No. Q8IW41
Gene Alias MAP kinase-activated protein kinase 5; MAPK-activated protein kinase 5; MAPKAP kinase 5; Mapkapk5; MAPKAP-K5; MAPKAPK-5; mitogen-activated protein kinase-activated protein kinase 5; MK5; MK-5; p38-regulated/activated protein kinase; PRAK
Gene Symbols MAPKAPK5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PRAK. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8550
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.