Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565403 Supplier Invitrogen™ Supplier No. PA565403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLPPMDPKLLK QLRKAEKAER EFRKKFKFEG EIVVHTKMMI DPNAKTRRGG GKHLGIRRGE ILEVIEFTSN EEMLCRDPKG KYGYVP Highest antigen sequence identity to the following orthologs - mouse 93%, rat 91%.

The protein encoded by this gene is similar to FYN binding protein (FYB/SLAP-130), an adaptor protein involved in T cell receptor mediated signaling. This gene is expressed and regulated during normal myelopoiesis. The expression of this gene is induced by retinoic acid and is inhibited by the expression of PML-RARalpha, a fusion protein of promyelocytic leukemia (PML) and the retinoic acid receptor-alpha (RARalpha).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAM1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRAM1
Gene Accession No. Q96QH2
Gene Alias AY665714; MGC39864; PML-RAR; PML-RAR alpha-regulated adaptor molecule 1; PML-RARA regulated adaptor molecule 1; PML-RARA target gene encoding an Adaptor Molecule-1; PML-RARA-regulated adapter molecule 1; PRAM; PRAM1; PRAM-1; PRAM-1 protein; RGD1564330
Gene Symbols PRAM1
Host Species Rabbit
Immunogen Recombinant Human PRAM1. Recombinant protein control fragment (Product #RP-106084).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84106
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.