Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAME Antibody (CL5148), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | PRAME |
---|---|
Clone | CL5148 |
Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:250 - 1:500, Immunohistochemistry-Paraffin 1:250 - 1:500 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PRAME Monoclonal specifically detects PRAME in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PRAME | |
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:250 - 1:500, Immunohistochemistry-Paraffin 1:250 - 1:500 | |
Unconjugated | |
Mouse | |
Human | |
PRAME | |
IgG1 | |
Protein A purified |
CL5148 | |
Monoclonal | |
Purified | |
Cancer | |
23532 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LLLSHIHASSYISPEKEEQYIAQFTSQFLSLQCLQALYVDSLFFLRGRLDQLLRHVMNPLETLSITNCRLSEGDVMHLSQSPSVSQLSVLSLSGVMLTDVSPEPLQALLERASATLQDLVFDECGI | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title