Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRAME Monoclonal Antibody (1E9G9), Proteintech
SDP

Catalog No. 502333079 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-3079 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-3079 Supplier Proteintech Group Inc Supplier No. 680971IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PRAME Monoclonal antibody specifically detects PRAME in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen PRAME
Applications Western Blot
Classification Monoclonal
Clone 1E9G9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. P78395
Gene Alias 4930534P07Rik; CT130; MAPE; OIP-4; OIP4; PRAME
Host Species Mouse
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23532
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.