Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAME Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583761
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583761 Supplier Invitrogen™ Supplier No. PA583761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLLSHIHASS YISPEKEEQY IAQFTSQFLS LQCLQALYVD SLFFLRGRLD QLLRHVMNPL ETLSITNCRL SEGDVMHLSQ SPSVSQLSVL SLSGVMLTDV SPEPLQALLE RASATLQDLV FDECGI Highest antigen sequence indentity to the following orthologs: Mouse - 55%, Rat - 55%.

This gene encodes an antigen that is predominantly expressed in human melanomas and that is recognized by cytolytic T lymphocytes. It is not expressed in normal tissues, except testis. This expression pattern is similar to that of other CT antigens, such as MAGE, BAGE and GAGE. However, unlike these other CT antigens, this gene is also expressed in acute leukemias. Five alternatively spliced transcript variants encoding the same protein have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAME
Applications Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRAME
Gene Accession No. P78395
Gene Alias 4930534P07Rik; cancer/testis antigen 130; CT130; MAPE; melanoma antigen preferentially expressed in tumors; OIP4; OIP-4; Opa-interacting protein 4; Opa-interacting protein OIP4; PRAME; preferentially expressed antigen in melanoma; preferentially expressed antigen of melanoma
Gene Symbols PRAME
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRAME. Recombinant protein control fragment (Product #RP-88805).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23532
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.