Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAMEF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116302
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116302 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116302 Supplier Invitrogen™ Supplier No. PA5116302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRAMEF1 Polyclonal Antibody for Western Blot

This gene is a member of the PRAME (preferentially expressed antigen of melanoma) gene family which is expressed in many cancers but may function in reproductive tissues during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAMEF1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PRAMEF1
Gene Accession No. O95521
Gene Alias PRAME family member 1; PRAMEF1
Gene Symbols PRAMEF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 427-454 amino acids from the C-terminal region of human PRAMEF1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65121
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.