Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAMEF18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598083 Supplier Invitrogen™ Supplier No. PA598083
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PRAMEF18 Polyclonal Antibody for Western Blot, IHC (P), ELISA

PRAMEF18 is a protein coding gene and gene ontology (GO) annotation includes cytoplasm; negative regulation of apoptotic process; negative regulation of cell differentiation; negative regulation of transcription, DNA-templated; positive regulation of cell population proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAMEF18
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PRAMEF18
Gene Accession No. Q5VWM3
Gene Alias PRAME family member 18; PRAME family member-like; PRAMEF18
Gene Symbols PRAMEF18
Host Species Rabbit
Immunogen Recombinant Human PRAME family member 18 protein (1-257AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 391003
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.