Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRAMEF20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548335
Description
PRAMEF20 Polyclonal Antibody for Western Blot

Specifications
PRAMEF20 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q5VT98 | |
PRAMEF20 | |
KLH conjugated synthetic peptide between 394-423 amino acids from the C-terminal region of human PRAMEF20. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PRAMEF20 | |
EG627009; Gm13125; novel preferentially expressed antigen in melanoma (Prame) protein; OTTHUMT00000008157; OTTMUSG00000010547; PRAME family member 20; PRAME family member 20/21; PRAME family member 21; PRAME family member 25-like; PRAMEF20; PRAMEF21; pramef25; predicted gene 13125; preferentially expressed antigen in melanoma-like 1-like; similar to PRAME family member 5 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
645425 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction