Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAMEF20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588672 Supplier Invitrogen™ Supplier No. PA588672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GALKKRTPYL ETQLQVVLEE VDKLLIQEVH PREYKLEVLD LRSVGQNYLN VWPGAMDDWL PKTTQTDPCC SKTVMTQPLK VLVDLQLNNR GQSRLFSYLV QWSDRRKGLL QLYCNKLQIW SPSQQNYRKL SQKVNLDYVE TLGLHSSCSP SFLLNFAPYL RQMRNLRKLA LSNIREEHII SPEKRRCIIT RFTLQILKLE SLQKLHLD; Positive Samples: Mouse skin, Rat heart.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAMEF20
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRAMEF20
Gene Accession No. Q5VT98
Gene Alias EG627009; Gm13125; novel preferentially expressed antigen in melanoma (Prame) protein; OTTHUMT00000008157; OTTMUSG00000010547; PRAME family member 20; PRAME family member 20/21; PRAME family member 21; PRAME family member 25-like; PRAMEF20; PRAMEF21; pramef25; predicted gene 13125; preferentially expressed antigen in melanoma-like 1-like; similar to PRAME family member 5
Gene Symbols PRAMEF20
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 67-274 of mouse GM13125 (NP_0011085491).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 627009, 645425, 691150
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.