Learn More
Invitrogen™ PRAMEF20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596928
Description
Immunogen sequence: GALKKRTPYL ETQLQVVLEE VDKLLIQEVH PREYKLEVLD LRSVGQNYLN VWPGAMDDWL PKTTQTDPCC SKTVMTQPLK VLVDLQLNNR GQSRLFSYLV QWSDRRKGLL QLYCNKLQIW SPSQQNYRKL SQKVNLDYVE TLGLHSSCSP SFLLNFAPYL RQMRNLRKLA LSNIREEHII SPEKRRCIIT RFTLQILKLE SLQKLHLD; Positive Samples: Mouse skeletal muscle, Rat heart.

Specifications
PRAMEF20 | |
Polyclonal | |
Unconjugated | |
PRAMEF20 | |
EG627009; Gm13125; novel preferentially expressed antigen in melanoma (Prame) protein; OTTHUMT00000008157; OTTMUSG00000010547; PRAME family member 20; PRAME family member 20/21; PRAME family member 21; PRAME family member 25-like; PRAMEF20; PRAMEF21; pramef25; predicted gene 13125; preferentially expressed antigen in melanoma-like 1-like; similar to PRAME family member 5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
627009, 645425, 691150 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.68 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5VT98 | |
PRAMEF20 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 67-274 of mouse Gm13125 (NP_0011085491). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.