Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PRAMEF6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571533
Description
PRAMEF6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Substrate-recognition component of a Cul2-RING (CRL2) E3 ubiquitin-protein ligase complex, which mediates ubiquitination of target proteins, leading to their degradation. The CRL2(PRAMEF6) complex mediates ubiquitination and degradation of truncated MSRB1/SEPX1 selenoproteins produced by failed UGA/Sec decoding.
Specifications
PRAMEF6 | |
Polyclonal | |
Unconjugated | |
PRAMEF6 | |
PRAME family member 6; PRAMEF6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
440561 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q5VXH4 | |
PRAMEF6 | |
KLH conjugated synthetic peptide between 395-423 amino acids from the C-terminal region of human PRAMEF6. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction