Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pramoxine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18605772
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum

PR106, 637-58-1, C17H27NO3·HCl

Spectrum™ Chemical Pramoxine Hydrochloride, USP is used as a topical anesthetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 637-58-1
Molecular Formula C17H28ClNO3
Molecular Weight (g/mol) 329.87
InChI Key SYCBXBCPLUFJID-UHFFFAOYSA-N
IUPAC Name 4-[3-(4-butoxyphenoxy)propyl]morpholine hydrochloride
SMILES Cl.CCCCOC1=CC=C(OCCCN2CCOCC2)C=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.01
Melting Range 170-174°C
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.