Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$201.00 - $261.00
Specifications
Antigen | PRAP1 |
---|---|
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PRAP1 Polyclonal antibody specifically detects PRAP1 in Human samples. It is validated for Western BlotSpecifications
PRAP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
PRO1195, proline-rich acidic protein 1, RP11-122K13.6, UPAMGC126792, Uterine-specific proline-rich acidic protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PRAP1. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1:500-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
118471 | |
IgG | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title