Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAP1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299413PCP
Description
PRAP1 Polyclonal antibody specifically detects PRAP1 in Human samples. It is validated for Western BlotSpecifications
PRAP1 | |
Polyclonal | |
Western Blot | |
PRO1195, proline-rich acidic protein 1, RP11-122K13.6, UPAMGC126792, Uterine-specific proline-rich acidic protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PRAP1. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
118471 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction