Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PRAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558508
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558508 Supplier Invitrogen™ Supplier No. PA558508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PILPGTKAWM ETEDTLGRVL SPEPDHDSLY HPPPEEDQGE ERPRLWVMPN HQVLLGPEED QDHIYH Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 46%.

Proline rich acidic protein 1 (PRAP1) is also named as PRO1195 or UPA. It may play an important role in maintaining normal growth homeostasis in epithelial cells. Mitotic arrest deficient 1 (MAD1) is a key component in mitotic checkpoint signaling. Recent study identified PRAP1 as a novel MAD1 binding partner using yeast-two hybrid screening. PRAP1 has four isoforms with MW 16-20 kDa and 10 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRAP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRAP1
Gene Accession No. Q96NZ9
Gene Alias epididymis tissue protein Li 178; PRAP1; Pregnancy-specific uterine protein; PRO1195; proline rich acidic protein 1; proline-rich acidic protein 1; UNQ608/PRO1195; Upa; uterine-specific proline-rich acidic protein
Gene Symbols PRAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRAP1. Recombinant protein control fragment (Product #RP-97157).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118471
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.