Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAS40 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PRAS40 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
PRAS40 Polyclonal specifically detects PRAS40 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PRAS40 | |
Unconjugated | |
RUO | |
Q96B36 | |
84335 | |
Synthetic peptides corresponding to AKT1S1(AKT1 substrate 1 (proline-rich)) The peptide sequence was selected from the C terminal of AKT1S1. Peptide sequence KSLPVSVPVWGFKEKRTEARSSDEENGPPSSPDLDRIAASMRALVLREAE. | |
Primary |
Polyclonal | |
Rabbit | |
Apoptosis, Autophagy, Cancer, Growth and Development, Lipid and Metabolism, mTOR Pathway, Neuronal Cell Markers, Neuroscience, Tumor Suppressors | |
40 kDa, AKT1 substrate 1 (proline-rich), MGC2865, proline-rich AKT1 substrate 1,40 kDa proline-rich AKT substrate | |
AKT1S1 | |
IgG | |
27 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title