Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRAS40 Antibody (SR1679), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PRAS40 |
---|---|
Clone | SR1679 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation |
Classification | Monoclonal |
Description
PRAS40 Monoclonal antibody specifically detects PRAS40 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunoprecipitationSpecifications
PRAS40 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
40 kDa, AKT1 substrate 1 (proline-rich), MGC2865, proline-rich AKT1 substrate 1,40 kDa proline-rich AKT substrate | |
A synthesized peptide derived from human PRAS40 (Uniprot #: Q96B36) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1679 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Autophagy, Cancer, Growth and Development, Lipid and Metabolism, mTOR Pathway, Neuronal Cell Markers, Neuroscience, Phospho Specific, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
84335 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title